Gene Transcripts

Gene Symbol
NICN1
Entrez Gene ID
84276
Gene Name
nicolin 1
Gene Aliases
Chromosome Location
Chr. 3: 49422333 - 49429324 on Build GRCh38
UniGene ID
Hs.191460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032316.3 NP_115692.1 2 231
GenBank mRNA AJ299740.1 - 2 203
AK094248.1 - 2 171
AK301846.1 - 2 234
AK313073.1 - 2 228
BC005050.1 - 2 148 2820022
BC050653.1 - 2 195 6015080

View Full Product Details