Gene Transcripts

Gene Symbol
PRADC1
Entrez Gene ID
84279
Gene Name
protease associated domain containing 1
Gene Aliases
C2orf7, PAP21
Chromosome Location
Chr. 2: 73228006 - 73233228 on Build GRCh38
UniGene ID
Hs.61268
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032319.1 NP_115695.1 4 450
GenBank mRNA AK312077.1 - 4 458
AY358605.1 - 4 441
BC005069.1 - 4 450 3530601

View Full Product Details