Gene Transcripts

Gene Symbol
RHOXF2
Entrez Gene ID
84528
Gene Name
Rhox homeobox family member 2
Gene Aliases
CT107, PEPP-2, PEPP2, THG1
Chromosome Location
Chr. X: 120158534 - 120164039 on Build GRCh38
UniGene ID
Hs.567620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032498.2 NP_115887.1 3, 4 733
XM_005262484.3 XP_005262541.1 3, 4 747
GenBank mRNA AF317219.1 - 3, 4 673
AK058125.1 - 3, 4 626
AL590526.1 - 3, 4 733
BC021719.2 - 3, 4 628 5269333

Gene Symbol
RHOXF2B
Entrez Gene ID
727940
Gene Name
Rhox homeobox family member 2B
Gene Aliases
Chromosome Location
Chr. X: 120072264 - 120077742 on Build GRCh38
UniGene ID
Hs.196956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099685.1 NP_001093155.1 3, 4 733
GenBank mRNA AL590524.1 - 3, 4 629

View Full Product Details