Gene Transcripts

Gene Symbol
GPAT3
Entrez Gene ID
84803
Gene Name
glycerol-3-phosphate acyltransferase 3
Gene Aliases
AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1
Chromosome Location
Chr. 4: 83535634 - 83605875 on Build GRCh38
UniGene ID
Hs.99196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256421.1 NP_001243350.1 10, 11 1209
NM_001256422.1 NP_001243351.1 10, 11 1213
NM_032717.4 NP_116106.2 9, 10 1296
XM_011532383.1 XP_011530685.1 8, 9 1102
XM_011532384.2 XP_011530686.1 10, 11 3297
XM_017008780.1 XP_016864269.1 10, 11 1365
XM_017008781.1 XP_016864270.1 10, 11 1372
GenBank mRNA AB075872.1 - 6, 7 494
AK055749.1 - 10, 11 1208
AK223543.1 - 10, 11 1180
AY358100.1 - 9, 10 1113
BC073136.1 - 4, 5 272
BC090956.1 - 9, 10 1202 30387839
DQ324782.1 - 9, 10 993
DQ345298.1 - 9, 10 1293

View Full Product Details