Gene Transcripts
- Gene Symbol
- KRTAP4-7
- Entrez Gene ID
- 100132476
- Gene Name
- keratin associated protein 4-7
- Gene Aliases
- KAP4.7, KRTAP4.7
- Chromosome Location
- Chr. 17: 41084207 - 41085144 on Build GRCh38
- UniGene ID
- Hs.632746
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033061.3 |
NP_149050.3
|
1 |
817 |
|
- Gene Symbol
- KRTAP4-9
- Entrez Gene ID
- 100132386
- Gene Name
- keratin associated protein 4-9
- Gene Aliases
- KAP4.9
- Chromosome Location
- Chr. 17: 41105389 - 41106488 on Build GRCh38
- UniGene ID
- Hs.307018
- Species
- Homo sapiens
View Full Product Details