Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033396.2 | NP_203754.2 | 6 | 3848 | |
XM_006718725.3 | XP_006718788.1 | 6 | 3864 | ||
XM_011545325.2 | XP_011543627.1 | 6 | 3861 | ||
GenBank mRNA | AB051528.2 | - | 6 | 3828 | |
AF441771.1 | - | 5 | 3990 | ||
AK074113.1 | - | 2 | 2205 | ||
AK124903.1 | - | 6 | 3875 | ||
AK127871.1 | - | 4 | 3271 | ||
BC150333.1 | - | 6 | 3839 | 8860508 |