Gene Transcripts
- Gene Symbol
- EPT1
- Entrez Gene ID
- 85465
- Gene Name
- ethanolaminephosphotransferase 1
- Gene Aliases
- SELI, SEPI
- Chromosome Location
- Chr. 2: 26346086 - 26395891 on Build GRCh38
- UniGene ID
- Hs.189073
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB051511.1 |
- |
8 |
913 |
|
AK095620.1 |
- |
8 |
923 |
|
View Full Product Details