Gene Transcripts

Gene Symbol
PLCZ1
Entrez Gene ID
89869
Gene Name
phospholipase C zeta 1
Gene Aliases
NYD-SP27, PLC-zeta-1, PLCzeta
Chromosome Location
Chr. 12: 18645529 - 18738059 on Build GRCh38
UniGene ID
Hs.743832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033123.3 NP_149114.2 11 1473
NR_073075.1 11 1476
XM_005253519.2 XP_005253576.1 10 1589
XM_011520883.1 XP_011519185.1 8 879
XM_017020176.1 XP_016875665.1 10 1589
XM_017020177.1 XP_016875666.1 11 1466
XM_017020178.1 XP_016875667.1 9 1439
XM_017020179.1 XP_016875668.1 10 1584
XM_017020180.1 XP_016875669.1 10 1478
XM_017020181.1 XP_016875670.1 10 1543
XM_017020182.1 XP_016875671.1 6 1242
XM_017020183.1 XP_016875672.1 7 1006
XM_017020184.1 XP_016875673.1 7 934
XM_017020185.1 XP_016875674.1 6 819
XM_017020186.1 XP_016875675.1 8 841
GenBank mRNA AF532185.1 - 11 1398
AK097555.1 - 7 821
AK097725.1 - 7 939
AK292279.1 - 11 1471
AK314516.1 - 11 1430
AY035866.1 - 11 1435
BC125067.1 - 7 726 40117397

View Full Product Details