Gene Transcripts

Gene Symbol
C19orf52
Entrez Gene ID
90580
Gene Name
chromosome 19 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 19: 10928748 - 10930241 on Build GRCh38
UniGene ID
Hs.164026
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138358.2 NP_612367.1 2 1344
GenBank mRNA BC005075.1 - 2 1321
BC007244.1 - 2 1334 2966591
BC011833.2 - 2 1344 3941883

View Full Product Details