Gene Transcripts
- Gene Symbol
- ANKRD44
- Entrez Gene ID
- 91526
- Gene Name
- ankyrin repeat domain 44
- Gene Aliases
- PP6-ARS-B
- Chromosome Location
- Chr. 2: 196962617 - 197310797 on Build GRCh38
- UniGene ID
- Hs.432706
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195144.1 |
NP_001182073.1
|
17, 18 |
1914 |
|
XM_005246945.1 |
XP_005247002.1
|
17, 18 |
1958 |
|
XM_005246946.1 |
XP_005247003.1
|
17, 18 |
1904 |
|
XM_005246947.2 |
XP_005247004.1
|
17, 18 |
1958 |
|
XM_005246948.2 |
XP_005247005.1
|
17, 18 |
1958 |
|
XM_006712838.3 |
XP_006712901.1
|
17, 18 |
1958 |
|
XM_011512148.2 |
XP_011510450.1
|
17, 18 |
1958 |
|
XM_017005252.1 |
XP_016860741.1
|
17, 18 |
1904 |
|
GenBank mRNA |
AK097086.1 |
- |
17, 18 |
1723 |
|
AL133087.1 |
- |
11, 12 |
1167 |
|
View Full Product Details