Gene Transcripts

Gene Symbol
RBM18
Entrez Gene ID
92400
Gene Name
RNA binding motif protein 18
Gene Aliases
Chromosome Location
Chr. 9: 122239555 - 122264864 on Build GRCh38
UniGene ID
Hs.415842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033117.3 NP_149108.1 3 379
NR_027125.1 3 379
NR_027126.1 4 473
XM_006717319.1 XP_006717382.1 3 335
GenBank mRNA AK057676.1 - 3 355
AK299625.1 - 4 449
AK310008.1 - 3 336
AK310807.1 - 4 415
AL389986.1 - 3 307
BC008942.2 - 3 326 2822634
BC019319.1 - 3 326 2822634
DA308339.1 - 3 379

View Full Product Details