Gene Transcripts

Gene Symbol
DIO1
Entrez Gene ID
1733
Gene Name
deiodinase, iodothyronine type I
Gene Aliases
5DI, TXDI1
Chromosome Location
Chr. 1: 53894187 - 53911086 on Build GRCh38
UniGene ID
Hs.251415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000792.6 NP_000783.2 1 133
NM_001039715.2 NP_001034804.1 1 133
NM_001039716.2 NP_001034805.1 1 133
NM_001324316.1 NP_001311245.1 1 133
NM_213593.4 NP_998758.1 1 133
NR_136692.1 1 133
NR_136693.1 1 133
GenBank mRNA AK290780.1 - 1 133
AY560374.1 - 1 109
AY560375.1 - 1 109
AY560376.1 - 1 109
AY560377.1 - 1 109
AY560378.1 - 1 109
AY560379.1 - 1 109
AY560381.1 - 1 109
AY560382.1 - 1 109
AY560383.1 - 1 109
BC017955.1 - 1 132 4607072
BC107170.2 - 1 132 40036824
BC107171.2 - 1 132 40036826
CD014030.1 - 1 115
FJ002243.1 - 1 131
FJ002244.1 - 1 131
FJ002245.1 - 1 131
FJ002247.1 - 1 131
FJ002248.1 - 1 131

View Full Product Details