Gene Transcripts

Gene Symbol
ZBTB47
Entrez Gene ID
92999
Gene Name
zinc finger and BTB domain containing 47
Gene Aliases
ZNF651
Chromosome Location
Chr. 3: 42653684 - 42667580 on Build GRCh38
UniGene ID
Hs.409561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145166.3 NP_660149.2 3 1804
GenBank mRNA AK127065.1 - 4 1498
AL133062.1 - 2 497
BC021855.1 - 2 462 4943271

View Full Product Details