Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142463.2 | NP_001135935.1 | 9 | 1080 | |
NM_001288747.1 | NP_001275676.1 | 9 | 1080 | ||
NM_001363.4 | NP_001354.1 | 9 | 1080 | ||
NR_110021.1 | 8 | 1557 | |||
NR_110022.1 | 8 | 1676 | |||
NR_110023.1 | 9 | 1450 | |||
GenBank mRNA | AF067008.1 | - | 9 | 916 | |
AK225077.1 | - | 9 | 944 | ||
AK307672.1 | - | 9 | 969 | ||
BC010015.2 | - | 9 | 882 | 4303933 | |
JF279874.1 | - | 9 | 903 | ||
KC954523.1 | - | 8 | 1380 | ||
KC954524.1 | - | 8 | 1499 | ||
KC954525.1 | - | 9 | 1273 | ||
U59151.1 | - | 9 | 936 |