Gene Transcripts

Gene Symbol
WDR89
Entrez Gene ID
112840
Gene Name
WD repeat domain 89
Gene Aliases
C14orf150, MSTP050
Chromosome Location
Chr. 14: 63597039 - 63641923 on Build GRCh38
UniGene ID
Hs.655666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008726.2 NP_001008726.1 4 1121
NM_001258272.1 NP_001245201.1 3 1517
NM_080666.3 NP_542397.1 3 1012
XM_011536383.2 XP_011534685.1 2 1051
XM_011536385.2 XP_011534687.1 4 1019
GenBank mRNA AF115513.1 - 3 944
AK095324.1 - 3 960
BC010698.1 - 2 904 3870073
BX248008.1 - 3 1001

View Full Product Details