Gene Transcripts

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
Chromosome Location
Chr. 6: 136231030 - 136253659 on Build GRCh38
UniGene ID
Hs.121536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099286.2 NP_001092756.1 3 523
NM_001318738.1 NP_001305667.1 4 335
NM_138419.4 NP_612428.2 3 235
XM_011535410.2 XP_011533712.1 4 587
XM_011535412.2 XP_011533714.1 2 328
XM_011535413.2 XP_011533715.1 2 3442
GenBank mRNA AK125758.1 - 4 292
AK292303.1 - 3 474
BC063688.1 - 3 188 4455541
BG831782.1 - 3 226
DB016393.1 - 3 499

View Full Product Details