Gene Transcripts

Gene Symbol
C1QTNF6
Entrez Gene ID
114904
Gene Name
C1q and tumor necrosis factor related protein 6
Gene Aliases
CTFP6, CTRP6, ZACRP6
Chromosome Location
Chr. 22: 37180166 - 37199400 on Build GRCh38
UniGene ID
Hs.22011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031910.3 NP_114116.3 3 695
NM_182486.1 NP_872292.1 3 695
XM_011529857.1 XP_011528159.1 4 3552
XM_017028569.1 XP_016884058.1 3 2844
XM_017028570.1 XP_016884059.1 5 1462
XM_017028571.1 XP_016884060.1 5 3200
XM_017028572.1 XP_016884061.1 5 3189
XM_017028573.1 XP_016884062.1 5 978
XM_017028574.1 XP_016884063.1 4 771
GenBank mRNA AF329842.1 - 3 633
AK128125.1 - 8 4047
AK226149.1 - 2 1218
AK314060.1 - 3 652
AY358374.1 - 5 832
BC020551.1 - 3 694 2900683
BU535692.1 - 2 281
CR456451.1 - 3 645

View Full Product Details