Gene Transcripts

Gene Symbol
WDR31
Entrez Gene ID
114987
Gene Name
WD repeat domain 31
Gene Aliases
Chromosome Location
Chr. 9: 113313222 - 113340340 on Build GRCh38
UniGene ID
Hs.133331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006615.2 NP_001006616.1 7 778
NM_001012361.3 NP_001012361.1 8 911
NM_145241.4 NP_660284.1 8 908
NR_134677.1 8 840
XM_011518194.2 XP_011516496.1 7 822
GenBank mRNA AK074891.1 - 7 692
AK093240.1 - 8 840
AK313843.1 - 8 850
AL833635.1 - 8 757
BC012352.1 - 8 840 4634445
BC022982.1 - 8 837

View Full Product Details