Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136050.2 | NP_001129522.1 | 2 | 413 | |
NM_138452.2 | NP_612461.1 | 2 | 405 | ||
GenBank mRNA | AF418205.1 | - | 2 | 349 | |
AK058159.1 | - | 2 | 405 | ||
AL833917.1 | - | 2 | 308 | ||
BC014057.2 | - | 2 | 319 | 3622913 | |
BC015943.1 | - | 2 | 216 | 3922816 | |
BX247980.1 | - | 2 | 329 | ||
DB272453.1 | - | 2 | 413 |