Gene Transcripts

Gene Symbol
RAB3C
Entrez Gene ID
115827
Gene Name
RAB3C, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 5: 58582188 - 58859394 on Build GRCh38
UniGene ID
Hs.4290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317915.1 NP_001304844.1 2 393
NM_138453.3 NP_612462.1 2 355
GenBank mRNA AY026936.1 - 2 313
BC013033.2 - 2 287 3534915
BX394661.2 - 2 365
DB485923.1 - 2 359

View Full Product Details