Gene Transcripts

Gene Symbol
OSR2
Entrez Gene ID
116039
Gene Name
odd-skipped related transciption factor 2
Gene Aliases
Chromosome Location
Chr. 8: 98944403 - 98952104 on Build GRCh38
UniGene ID
Hs.253247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142462.2 NP_001135934.1 3 1249
NM_001286841.1 NP_001273770.1 4 1498
NM_053001.3 NP_443727.2 3 1249
XM_005250778.4 XP_005250835.1 4 1457
XM_011516825.2 XP_011515127.1 4 1264
XM_011516826.2 XP_011515128.1 3 890
XM_011516827.2 XP_011515129.1 3 997
XM_017013018.1 XP_016868507.1 3 869
GenBank mRNA AK074518.1 - 3 1210
AK291491.1 - 3 1210
AK304654.1 - 4 1242
AK310994.1 - 3 1210
AY038072.1 - 3 1029
AY038073.1 - 3 1211
BC016936.1 - 3 1195 4425397

View Full Product Details