Gene Transcripts

Gene Symbol
SLC26A8
Entrez Gene ID
116369
Gene Name
solute carrier family 26 member 8
Gene Aliases
SPGF3, TAT1
Chromosome Location
Chr. 6: 35943514 - 36024855 on Build GRCh38
UniGene ID
Hs.435836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193476.1 NP_001180405.1 17 2112
NM_052961.3 NP_443193.1 17 2136
NM_138718.2 NP_619732.2 15 1859
XM_011514294.2 XP_011512596.1 16 2277
XM_017010235.1 XP_016865724.1 17 2162
GenBank mRNA AF331522.1 - 17 2080
AK055314.1 - 7 1582
AK057276.1 - 7 655
AK097410.1 - 11 1198
AK122928.1 - 17 2112
BC025408.2 - 15 1852 4823098

View Full Product Details