Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145794.1 | NP_001139266.1 | 8 | 1414 | |
NM_001286780.1 | NP_001273709.1 | 8 | 577 | ||
NM_001286781.1 | NP_001273710.1 | 8 | 1114 | ||
NM_058172.5 | NP_477520.2 | 8 | 1414 | ||
XM_011531587.2 | XP_011529889.1 | 8 | 577 | ||
XM_017007711.1 | XP_016863200.1 | 8 | 1114 | ||
GenBank mRNA | AB209913.1 | - | 8 | 1002 | |
AK055636.1 | - | 7 | 465 | ||
AL832851.1 | - | 4 | 245 | ||
AY233452.1 | - | 8 | 654 | ||
BC107876.1 | - | 8 | 580 | 6108432 | |
BC114374.1 | - | 8 | 526 |