Gene Transcripts

Gene Symbol
GPR62
Entrez Gene ID
118442
Gene Name
G protein-coupled receptor 62
Gene Aliases
GPCR8, KPG_005
Chromosome Location
Chr. 3: 51955314 - 51957504 on Build GRCh38
UniGene ID
Hs.232213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080865.3 NP_543141.3 1 1707
GenBank mRNA BC026357.1 - 1 1640

View Full Product Details