Gene Transcripts

Gene Symbol
OR51S1
Entrez Gene ID
119692
Gene Name
olfactory receptor family 51 subfamily S member 1
Gene Aliases
OR11-24
Chromosome Location
Chr. 11: 4848237 - 4849208 on Build GRCh38
UniGene ID
Hs.553561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004758.1 NP_001004758.1 1 844
GenBank mRNA BC136932.1 - 1 920
BC136934.1 - 1 920 9020926

View Full Product Details