Gene Transcripts
- Gene Symbol
- LRR1
- Entrez Gene ID
- 122769
- Gene Name
- leucine rich repeat protein 1
- Gene Aliases
- 4-1BBLRR, LRR-1, PPIL5
- Chromosome Location
- Chr. 14: 49598697 - 49614672 on Build GRCh38
- UniGene ID
- Hs.451090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152329.3 |
NP_689542.2
|
1, 2 |
508 |
|
NM_203467.1 |
NP_982292.1
|
1, 2 |
508 |
|
NR_037792.1 |
|
1, 2 |
508 |
|
GenBank mRNA |
AK293156.1 |
- |
1, 2 |
227 |
|
AY052405.1 |
- |
1, 2 |
194 |
|
BC030142.2 |
- |
1, 2 |
444 |
|
BC093697.1 |
- |
1, 2 |
210 |
|
BC107888.1 |
- |
1, 2 |
257 |
|
BC112241.1 |
- |
1, 2 |
210 |
|
BC139921.1 |
- |
1, 2 |
248 |
40148567 |
BI825814.1 |
- |
1, 2 |
435 |
|
BI826606.1 |
- |
1, 2 |
509 |
|
BI829295.1 |
- |
1, 2 |
445 |
|
BQ059807.1 |
- |
1, 2 |
178 |
|
BX248298.1 |
- |
1, 2 |
251 |
|
View Full Product Details