Gene Transcripts

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
Chromosome Location
Chr. 14: 51489100 - 51730727 on Build GRCh38
UniGene ID
Hs.434914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042481.2 NP_001035946.1 14, 15 2077
NM_001267046.1 NP_001253975.1 13, 14 1877
NM_001267047.1 NP_001253976.1 4, 5 634
NM_152330.3 NP_689543.1 13, 14 1853
XM_006720029.1 XP_006720092.1 13, 14 1974
XM_006720030.1 XP_006720093.1 13, 14 1950
XM_011536423.1 XP_011534725.1 14, 15 2121
XM_011536424.1 XP_011534726.1 15, 16 2246
GenBank mRNA AK055545.1 - 13, 14 1788
AK126521.1 - 11, 12 1498
AK315764.1 - 13, 14 1774
AL833158.1 - 14, 15 2063
AL833380.1 - 8, 9 1092
AL833661.1 - 8, 9 1073
BC020521.2 - 13, 14 1801 3914276
BC029870.1 - 4, 5 634
BX161430.1 - 13, 14 1853
BX647239.1 - 4, 5 2422
BX647863.1 - 13, 14 1765
BX648295.1 - 4, 5 2423

View Full Product Details