Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173483.3 | NP_775754.2 | 8 | 861 | |
XM_011527692.2 | XP_011525994.1 | 9 | 1103 | ||
XM_011527693.2 | XP_011525995.1 | 8 | 873 | ||
GenBank mRNA | AK096820.1 | - | 8 | 861 | |
BC069351.1 | - | 7 | 798 | 7262372 | |
BC093894.1 | - | 7 | 735 | 7939739 | |
BC093896.1 | - | 7 | 735 | 7939741 |