Gene Transcripts

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
Chromosome Location
Chr. 1: 94820342 - 94895247 on Build GRCh38
UniGene ID
Hs.483423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114106.2 NP_001107578.1 8 918
NM_001258340.1 NP_001245269.1 8 918
NM_001258341.1 NP_001245270.1 8 822
NM_001258342.1 NP_001245271.1 7 810
NM_001258343.1 NP_001245272.1 7 714
NM_001301079.1 NP_001288008.1 7 757
NM_152369.4 NP_689582.2 7 853
XM_005270440.2 XP_005270497.1 8 813
XM_005270441.2 XP_005270498.1 8 855
XM_005270442.1 XP_005270499.1 8 909
XM_011540632.2 XP_011538934.1 7 782
XM_011540633.2 XP_011538935.1 7 620
XM_011540635.2 XP_011538937.1 8 909
XM_011540636.2 XP_011538938.1 8 813
XM_017000262.1 XP_016855751.1 7 795
XM_017000263.1 XP_016855752.1 8 855
XM_017000264.1 XP_016855753.1 8 909
XM_017000265.1 XP_016855754.1 8 813
XM_017000266.1 XP_016855755.1 8 909
GenBank mRNA AK300193.1 - 7 651
AK301288.1 - 7 659
AK302381.1 - 5 502
AK303906.1 - 7 789
AK313574.1 - 8 806
AK316123.1 - 9 763
AL530322.3 - 8 910
AL540829.3 - 7 810
AY358659.1 - 4 372
BC033858.1 - 8 881
BC053877.1 - 7 776 5760394
BX648225.1 - 7 782

View Full Product Details