Gene Transcripts

Gene Symbol
OR2T11
Entrez Gene ID
127077
Gene Name
olfactory receptor family 2 subfamily T member 11 (gene/pseudogene)
Gene Aliases
OR2T11Q
Chromosome Location
Chr. 1: 248626178 - 248627128 on Build GRCh38
UniGene ID
Hs.626619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001964.1 NP_001001964.1 1 655
GenBank mRNA BC140749.1 - 1 717

View Full Product Details