Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 7 774
NM_001287585.1 NP_001274514.1 7 794
NM_138285.4 NP_612142.2 6 671
NR_109856.1 5 529
XM_006712254.2 XP_006712317.1 6 636
XM_011510576.2 XP_011508878.1 8 902
XM_011510577.2 XP_011508879.1 8 1089
XM_017003308.1 XP_016858797.1 6 827
GenBank mRNA AF411516.1 - 6 671
AF514993.1 - 6 628
AK289601.1 - 5 472
AK290760.1 - 6 583
AK298199.1 - 7 704
AK302350.1 - 7 743
BC035597.1 - 6 595
BC047029.1 - 6 658 5541023
BC061698.1 - 6 625 4132440
BI821231.1 - 5 465

View Full Product Details