Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039569.1 | NP_001034658.1 | 4 | 499 | |
NR_110905.1 | 5 | 538 | |||
NR_110906.1 | 3 | 390 | |||
GenBank mRNA | AF393369.1 | - | 4 | 381 | |
AK025338.1 | - | 3 | 358 | ||
AK299026.1 | - | 4 | 479 | ||
BC009606.1 | - | 4 | 500 | 4284350 | |
BG340480.1 | - | 3 | 261 |