Gene Transcripts

Gene Symbol
FAM43A
Entrez Gene ID
131583
Gene Name
family with sequence similarity 43 member A
Gene Aliases
Chromosome Location
Chr. 3: 194685893 - 194689037 on Build GRCh38
UniGene ID
Hs.435080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153690.4 NP_710157.2 1 2841
GenBank mRNA BI494721.1 - 1 50

View Full Product Details