Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172700.1 | NP_001166171.1 | 7 | 2197 | |
NM_133456.2 | NP_597713.2 | 4 | 1562 | ||
XM_005271885.4 | XP_005271942.1 | 5 | 2563 | ||
XM_005271886.3 | XP_005271943.1 | 5 | 1475 | ||
XM_011543167.2 | XP_011541469.1 | 6 | 1957 | ||
GenBank mRNA | AB075840.1 | - | 7 | 2197 | |
AF314142.1 | - | 4 | 1562 | ||
BC104914.1 | - | 4 | 1424 | 8143917 | |
BC104916.1 | - | 4 | 1424 | 8143919 | |
BC143494.1 | - | 4 | 1424 | 9052002 |