Gene Transcripts

Gene Symbol
PAGE3
Entrez Gene ID
139793
Gene Name
PAGE family member 3
Gene Aliases
CT16.6, GAGED1, PAGE-3
Chromosome Location
Chr. X: 55257833 - 55264916 on Build GRCh38
UniGene ID
Hs.163473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017931.2 NP_001017931.2 4 525
NM_001171252.1 NP_001164723.1 4 539
NM_001303613.1 NP_001290542.1 4 327
XM_017029282.1 XP_016884771.1 4 328
GenBank mRNA BC140001.1 - 4 398
BC140741.1 - 4 411 9021609
BC144449.1 - 4 424 9052975
HY049011.1 - 4 327

View Full Product Details