Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012428.2 | NP_001012428.1 | 6 | 769 | |
NM_001201583.1 | NP_001188512.1 | 6 | 727 | ||
NM_080873.2 | NP_543149.1 | 6 | 778 | ||
GenBank mRNA | AF425642.1 | - | 6 | 759 | |
BC069340.1 | - | 6 | 759 | 7262279 | |
BC103873.1 | - | 7 | 877 | ||
BC103874.1 | - | 6 | 759 | 40004166 | |
BC103875.1 | - | 6 | 759 | 40004167 | |
BX537853.1 | - | 6 | 769 | ||
BX537857.1 | - | 6 | 727 |