Gene Transcripts

Gene Symbol
CBLN4
Entrez Gene ID
140689
Gene Name
cerebellin 4 precursor
Gene Aliases
CBLNL1
Chromosome Location
Chr. 20: 55997357 - 56005472 on Build GRCh38
UniGene ID
Hs.126141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080617.5 NP_542184.1 2, 3 1710
GenBank mRNA AK289640.1 - 2, 3 1514
AY358527.1 - 2, 3 745
BC050026.1 - 2, 3 1712 4820943
BC069402.1 - 2, 3 409 7262170
BC069488.1 - 2, 3 409 7262182
BC069516.1 - 2, 3 409 7262099

View Full Product Details