Gene Transcripts
- Gene Symbol
- FRMPD2B
- Entrez Gene ID
- 728798
- Gene Name
- FERM and PDZ domain containing 2B, pseudogene
- Gene Aliases
- FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2
- Chromosome Location
- Chr. 10: 46876674 - 46895326 on Build GRCh38
- UniGene ID
- Hs.664786
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033172.1 |
|
2, 3 |
484 |
|
GenBank mRNA |
AK294650.1 |
- |
2, 3 |
439 |
|
AK313659.1 |
- |
2, 3 |
441 |
|
AK315971.1 |
- |
2, 3 |
484 |
|
BC071635.1 |
- |
2, 3 |
396 |
|
BC072395.1 |
- |
2, 3 |
410 |
|
BC110798.1 |
- |
2, 3 |
407 |
|
BC137431.1 |
- |
2, 3 |
371 |
9021429 |
BC137434.1 |
- |
2, 3 |
371 |
9021432 |
BC137439.1 |
- |
2, 3 |
369 |
9021437 |
BC137440.1 |
- |
2, 3 |
371 |
9021438 |
BC144635.1 |
- |
2, 3 |
371 |
9053166 |
- Gene Symbol
- FRMPD2
- Entrez Gene ID
- 143162
- Gene Name
- FERM and PDZ domain containing 2
- Gene Aliases
- PDZD5C, PDZK4, PDZK5C
- Chromosome Location
- Chr. 10: 48156559 - 48274898 on Build GRCh38
- UniGene ID
- Hs.664786
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001018071.3 |
NP_001018081.3
|
25, 26 |
3562 |
|
NM_001042512.2 |
NP_001035977.2
|
2, 3 |
430 |
|
NM_001318191.1 |
NP_001305120.1
|
23, 24 |
3203 |
|
XM_017015744.1 |
XP_016871233.1
|
2, 3 |
246 |
|
GenBank mRNA |
AK123038.1 |
- |
23, 24 |
3203 |
|
BC031614.1 |
- |
4, 5 |
425 |
|
BC073954.1 |
- |
2, 3 |
411 |
|
BC144636.1 |
- |
2, 3 |
371 |
9053167 |
BC144639.1 |
- |
2, 3 |
371 |
9053170 |
CR749241.1 |
- |
25, 26 |
3534 |
|
View Full Product Details