Gene Transcripts

Gene Symbol
CCNYL1
Entrez Gene ID
151195
Gene Name
cyclin Y like 1
Gene Aliases
Chromosome Location
Chr. 2: 207711540 - 207756174 on Build GRCh38
UniGene ID
Hs.471234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152523.2 NP_689736.1 5, 6 544
XM_005246343.1 XP_005246400.1 4, 5 562
GenBank mRNA AK097751.1 - 5, 6 544

View Full Product Details