Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144715.3 | NP_653316.3 | 7 | 1631 | |
XM_005264889.1 | XP_005264946.1 | 7 | 1631 | ||
XM_011533382.1 | XP_011531684.1 | 6 | 1568 | ||
XM_011533383.2 | XP_011531685.2 | 7 | 739 | ||
XM_011533385.2 | XP_011531687.1 | 8 | 1007 | ||
XM_017005741.1 | XP_016861230.1 | 9 | 1191 | ||
XM_017005743.1 | XP_016861232.1 | 8 | 958 | ||
XM_017005744.1 | XP_016861233.1 | 7 | 870 | ||
XM_017005745.1 | XP_016861234.1 | 8 | 933 | ||
GenBank mRNA | AK097644.1 | - | 7 | 1633 | |
AK122616.1 | - | 9 | 1167 | ||
BC028198.1 | - | 7 | 1597 | 5209454 |