Gene Transcripts

Gene Symbol
ROPN1B
Entrez Gene ID
152015
Gene Name
rhophilin associated tail protein 1B
Gene Aliases
Chromosome Location
Chr. 3: 125969144 - 125983454 on Build GRCh38
UniGene ID
Hs.663128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012337.2 NP_001012337.1 5 829
NM_001308313.1 NP_001295242.1 6 875
XM_005247138.3 XP_005247195.1 4 684
XM_006713513.3 XP_006713576.1 7 971
GenBank mRNA AF231410.1 - 5 788
BC015413.1 - 3 569 4394326
BC141849.1 - 6 822 8860347

View Full Product Details