Gene Transcripts

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
Chromosome Location
Chr. 4: 158893532 - 159082883 on Build GRCh38
UniGene ID
Hs.415576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152543.2 NP_689756.2 4 592
XM_011531671.2 XP_011529973.1 5 627
XM_011531672.2 XP_011529974.1 5 737
XM_017007808.1 XP_016863297.1 8 1493
XM_017007809.1 XP_016863298.1 8 1423
XM_017007810.1 XP_016863299.1 8 1599
GenBank mRNA AK058100.1 - 4 592
AK307319.1 - 3 294
DB037602.1 - 3 294

View Full Product Details