Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130678.1 | NP_001124150.1 | 5 | 527 | |
NM_001130679.1 | NP_001124151.1 | 5 | 1870 | ||
NM_001968.3 | NP_001959.1 | 5 | 1870 | ||
XM_006714126.3 | XP_006714189.1 | 6 | 1573 | ||
GenBank mRNA | AK295083.1 | - | 5 | 363 | |
AK300982.1 | - | 5 | 527 | ||
BC012611.1 | - | 5 | 363 | 4282826 | |
BC035166.1 | - | 5 | 1878 | 5265600 | |
BC043226.1 | - | 5 | 1209 | 5295521 | |
BC107715.1 | - | 5 | 364 | 6716042 | |
BM849222.1 | - | 5 | 361 | ||
DC388666.1 | - | 5 | 527 | ||
KJ534830.1 | - | 5 | 407 | ||
M15353.1 | - | 5 | 365 |