Gene Transcripts

Gene Symbol
TAS2R30
Entrez Gene ID
259293
Gene Name
taste 2 receptor member 30
Gene Aliases
T2R30, T2R47, TAS2R47
Chromosome Location
Chr. 12: 11133285 - 11134244 on Build GRCh38
UniGene ID
Hs.679464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097643.1 NP_001091112.1 1 477

View Full Product Details