Gene Transcripts

Gene Symbol
CLEC2L
Entrez Gene ID
154790
Gene Name
C-type lectin domain family 2 member L
Gene Aliases
Chromosome Location
Chr. 7: 139523928 - 139544985 on Build GRCh38
UniGene ID
Hs.57806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080511.2 NP_001073980.2 3, 4 435
XM_017011770.1 XP_016867259.1 3, 4 386

View Full Product Details