Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001419.2 | NP_001410.2 | 3 | 372 | |
GenBank mRNA | AK301013.1 | - | 3 | 374 | |
BC003376.2 | - | 3 | 335 | 2901220 | |
BT009793.1 | - | 2 | 205 | ||
BU542524.1 | - | 3 | 373 | ||
U38175.1 | - | 3 | 323 |