Gene Transcripts

Gene Symbol
ITPRIPL2
Entrez Gene ID
162073
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 2
Gene Aliases
Chromosome Location
Chr. 16: 19113932 - 19121630 on Build GRCh38
UniGene ID
Hs.530899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034841.3 NP_001030013.1 1 2639
NR_028028.1 2 2123

View Full Product Details