Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145545.1 | NP_001139017.1 | 1 | 441 | |
GenBank mRNA | AF536240.1 | - | 1 | 632 | |
AK311574.1 | - | 1 | 383 | ||
BC031257.1 | - | 1 | 414 | ||
BC136370.1 | - | 1 | 624 | 9020357 | |
BC136382.1 | - | 1 | 624 | 9020369 |