Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166002.2 | NP_001159474.2 | 4 | 589 | |
NM_001166003.2 | NP_001159475.2 | 4 | 589 | ||
NM_181773.4 | NP_861438.3 | 4 | 589 | ||
XM_011529990.2 | XP_011528292.1 | 4 | 609 | ||
XM_011529991.2 | XP_011528293.1 | 4 | 609 | ||
XM_011529992.2 | XP_011528294.1 | 4 | 608 | ||
GenBank mRNA | BC069023.1 | - | 4 | 574 | |
BQ052182.1 | - | 4 | 528 | ||
BQ054330.1 | - | 4 | 535 | ||
CR456481.3 | - | 4 | 502 | ||
FJ376611.1 | - | 3 | 462 | ||
FJ376612.1 | - | 3 | 462 | ||
FJ376613.1 | - | 3 | 462 | ||
FJ376614.1 | - | 3 | 462 | ||
FJ376615.1 | - | 3 | 462 | ||
FJ376616.1 | - | 3 | 462 | ||
KC329529.1 | - | 3 | 462 |