Gene Transcripts
- Gene Symbol
- SLC29A1
- Entrez Gene ID
- 2030
- Gene Name
- solute carrier family 29 member 1 (Augustine blood group)
- Gene Aliases
- ENT1
- Chromosome Location
- Chr. 6: 44219505 - 44234151 on Build GRCh38
- UniGene ID
- Hs.25450
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001078175.2 |
NP_001071643.1
|
6, 7 |
742 |
|
NM_001078177.1 |
NP_001071645.1
|
6, 7 |
937 |
|
NM_001304462.1 |
NP_001291391.1
|
6, 7 |
875 |
|
NM_001304463.1 |
NP_001291392.1
|
5, 6 |
635 |
|
NM_001304465.1 |
NP_001291394.1
|
5, 6 |
778 |
|
NM_001304466.1 |
NP_001291395.1
|
5, 6 |
775 |
|
XM_005248876.4 |
XP_005248933.1
|
5, 6 |
600 |
|
XM_005248878.3 |
XP_005248935.1
|
6, 7 |
772 |
|
XM_005248879.3 |
XP_005248936.1
|
6, 7 |
706 |
|
XM_005248880.3 |
XP_005248937.1
|
6, 7 |
683 |
|
XM_005248881.3 |
XP_005248938.1
|
5, 6 |
780 |
|
XM_005248882.3 |
XP_005248939.1
|
5, 6 |
723 |
|
XM_011514341.2 |
XP_011512643.1
|
6, 7 |
796 |
|
GenBank mRNA |
AF079117.1 |
- |
5, 6 |
548 |
|
AK090491.1 |
- |
6, 7 |
744 |
|
AK090615.1 |
- |
6, 7 |
763 |
|
AX775997.1 |
- |
5, 6 |
624 |
|
BC001382.2 |
- |
5, 6 |
548 |
3051441 |
BC008954.2 |
- |
6, 7 |
621 |
3010092 |
U81375.1 |
- |
5, 6 |
624 |
|
View Full Product Details